Learn More
Corning™ 96-Well Filter Plate Fluid Guard
Thermoformed polystyrene fluid guard
Brand: Corning™ 3514
Code : 66
Additional Details : Weight : 0.77111kg Transport : UN number : 1684 Chem class : 6.1 Pack group : II
Description
The fluid guard is manufactured from thermoformed polystyrene, non-sterile and easily snaps to the base of the filter plate. The unique design enables the guard to be applied manually or robotically to the plate. The guard serves to protect laboratory furniture and equipment from hazardous or radioactive materials.Specifications
Standard | |
Fluid Guard | |
96 | |
96-well filter plates | |
100/Pk. |
8 x 12 | |
Polystyrene | |
Non-sterile | |
100/Cs. |