missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Flat-Bottom Polystyrene NBS Microplate Without Lid

Product Code. 10656853 Shop All Corning Products
Change view
Click to view available options
Color:
Black
White
Volume (Metric) Working:
25 to 125 μL
75 to 200 μL
Format:
Half-Area
Standard
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Working Format Unit Size
10656853 Black 75 to 200 μL Standard Pack of 25
10173682 White 25 to 125 μL Half-Area Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656853 Supplier Corning™ Supplier No. 3991
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    306.28 € / Pack of 25
    Save 21.72 €
    6% Off
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nonsterile

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color Black
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 25/Cs.
    No. per Pack 5/Pk.
    Quantity 25/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.