Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. 16485399 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Product Code. Color unitSize
16485399 Clear Pack of 100
1 options
Product Code. 16485399 Supplier Corning™ Supplier No. 3628BC
Only null left
Add to basket
Add to basket
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Packaging 50/Pk., 100/Cs.
Binding Type TC-Treated
Color Clear
Product Type Assay Plate
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 100/Cs.