missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Half Area Flat Bottom Polystyrene NBS™ Microplates
GREENER_CHOICE

Product Code. 10522063 Shop All Corning Products
Change view
Click to view available options
Color:
White
White, Clear Bottom
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10522063 White Pack of 100
10317722 White, Clear Bottom Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10522063 Supplier Corning™ Supplier No. 3642
  • ONLINE EXCLUSIVE PRICE
    1245.00 €
    1163.45 € / Pack of 100
    Save 81.55 €
    6% Off
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to reduce well-to-well crosstalk during luminescent assays

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Half-Area
    Lid Without Lid
    Array 8 x 12
    Color White
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.