missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Laminin/Fibronectin Multiwell Plate
Description
Useful for construction of neural cell model systems to study central nervous system (CNS) function, development and diseases—as well as for the discovery of new therapeutic agents.
- Both laminin (LM) and human fibronectin (HFN) have been shown to be involved in neuronal cell adhesion, neural crest cell migration, neurite outgrowth, growth and regeneration of axons
- Human fibronectin is derived from human plasma; laminin from EHS mouse tumor
- Primary rat cortical neuron enriched cultures are tested for receptor agonist induced changes in intracellular calcium using Fluo-3A (long-wavelength fluorescent calcium indicator)
- Each lot has been tested and found negative for bacteria and fungi
Human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody.
Specifications
Specifications
| Format | Standard |
| Content And Storage | 2°C to 8°C |
| Culture Area | 0.32 cm2 |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Surface Treatment | Cell Culture |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?