Learn More
Corning™ 96-Well, CellBIND-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. 10386612 Shop All Corning Products
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 50
Product Code. 10386612
Brand: Corning™ 3340
Only null left
Add to basket
Add to basket
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid With Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Color Black, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment CellBIND
    Total Volume (Metric) Well 360 μL
    Well Shape Flat Bottom (F-well)
    For Use With (Application) CellBIND-treated surface helps cells adapt to reduced serum or serum-free media and improves cell attachment for increased cell growth and yield. Requires no refrigeration or special handling, unlike standard coatings used to improve cell attachment.
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Quantity 50/Cs.