Learn More
Corning™ CellBIND™ 96-Well, CellBIND-Treated, Flat-Bottom Microplate
BIOPHARMAGREENER_CHOICE
Product Code. 10510733 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
Product Code. Color Unit Size
10510733 Clear Pack of 50
1 options
Product Code. 10510733 Supplier Corning™ Supplier No. 3300
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
In Stock
Add to basket
Edge
Add to basket
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type Corning CellBIND Surface
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment CellBIND
Total Volume (Metric) Well 360 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) CellBIND-treated surface helps cells adapt to reduced serum or serum-free media and improves cell attachment for increased cell growth and yield. Requires no refrigeration or special handling, unlike standard coatings used to improve cell attachment.
No. per Case 50/Cs.
No. per Pack 5/Pk.
Quantity 50/Cs.
Well Color Clear