Learn More
Corning™ CellBIND™ Surface Microplates
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
229.00 € - 322.00 €
Specifications
Closure Included | Yes |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Surface Treatment | CellBIND |
No. per Case | 50/Cs. |
Product Code | Brand | Culture Area | Volume (Metric) Working | Well Count | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Volume (Metric) Working | Well Count | Capacity (Metric) | Price | Quantity & Availability | ||
10739864
|
Corning™
3336 |
3.8 cm2 | 0.76 to 1.14 mL | 12 | 6.9 mL |
264.00 €
Pack of 50 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||
10234832
|
Corning™
3335 |
9.5 cm2 | 1.9 to 2.9 mL | 6 | 2.9 mL (Well) |
229.00 €
Pack of 50 |
Estimated Shipment: 23-04-2024
Log in to see stock. |
||
10224882
|
Corning™
3337 |
1.9 cm2 | 0.38 to 0.57 mL | 24 | 0.57 mL (Well) |
239.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||
10251443
|
Corning™
3338 |
0.95 cm2 | 0.19 to 0.28 mL | 48 | 0.28 mL (Well) |
322.00 €
Pack of 50 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||
Description
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Certifications
Certified nonpyrogenic
Specifications
Yes | |
Sterile | |
50/Cs. |
Polystyrene | |
CellBIND | |
5/Pk. |