missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This 50L gusseted tank liner is designed to eliminate cross contamination and the need to clean and validate tanks, while extending reusable tank life. Each bag is irradiated and individually packed to ensure sterility. It is ideal for biopharmaceutical applications.
Corning’s portfolio of irradiated tank liners are designed to fit cylindrical tanks. Tank liners are manufactured with low-density polyethylene (ULDPE) and are available in a range of sizes. Features and Benefits
Gamma irradiated, individually packaged Free of animal-derived components Eliminate costs associated with the cleaning and validation of tanks Decrease labor costs and increase turnaround time Eliminate the risks associated with cross contamination Help extend the life of reusable tanks, resulting in lower capital expenditures Wide variety of sizes available
Corning media certificates are available on our Resource Library.
Corning’s portfolio of irradiated tank liners are designed to fit cylindrical tanks. Tank liners are manufactured with low-density polyethylene (ULDPE) and are available in a range of sizes. Features and Benefits
Corning media certificates are available on our Resource Library.
Specifications
Specifications
| Capacity (Metric) | 50 L |
| Material | LDPE |
| Capacity (English) | 0.0132 gal. |
| For Use With (Application) | Ideal for Biopharmaceutical Application |
| Product Type | Gusseted 3D Tank Liner |
Product Title
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?