missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix
BIOPHARMA

Product Code. 15373541
Change view
Click to view available options
Quantity:
6 x 500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15373541 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15373541 Supplier Corning™ Supplier No. 10090CV
  • ONLINE EXCLUSIVE PRICE
    83.40 €
    36.64 € / Pack of 6
    Save 46.76 €
    56% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for culture of epithelial, endothelial, and granulosa cell types under low serum conditions. Corning™ cellgro™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix has low levels of endotoxin, and absence of contamination.

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Fifty-fifty mixture of DMEM and F-12, originally formulated for rat neuroblastoma cells and MDCK cells
  • Serum-free growth may be possible with appropriate protein supplements
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Formulation of DMEM/Ham's F-12 contains L-glutamine
  • Sterile
TRUSTED_SUSTAINABILITY

Especificaciones

Concentration 1 X
Form Liquid
Product Type DMEM and Ham's F-12, 50/50 Mix
Sterility Sterile
With Additives Glutamine
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 500 mL
Endotoxin Level <0.25EU/mL
Formulation Inorganic Salts: 116.65mg/L CaCl2 (anhydrous), 0.417mg/L FeSO4.7H2O, 0.05mg/L Fe(NO3)3.9H2O, 311.8mg/L KCl, 0.0008mg/L CuSO4 (anhydrous), 84.95mg/L MgSO4 (anhydrous), 7000mg/L NaCl, 62.5mg/L Na2HPO4.H2O, 71mg/L Na2HPO4 (anhydrous), 2438mg/L NaHCO3, 0.4315mg/L ZnSO4.7H2O; Amino Acids: 4.45mg/L L-Alanine, 147.5mg/L L-Arginine. HCl, 7.5mg/L L-Asparagine.H2O, 6.65mg/L L-Aspartic acid, 17.56mg/L L-Cysteine.HCl.H2O, 31.285mg/L L-Cysteine.2HCl, 7.35mg/L L-Glutamic acid, 365.1mg/L L-glutamine, 18.75mg/L Glycine, 31.48mg/L L-Histidine.HCl.H2O, 54.37mg/L L-Isoleucine, 58.95mg/L L-Leucine, 91.35mg/L L-Lysine. HCl, 17.24mg/L L-Methionine, 35.48mg/L L-Phenylalanine, 17.25mg/L L-Proline, 26.25mg/L L-Serine, 53.55mg/L L-Threonine, 9.02mg/L L-Tryptophan, 55.815mg/L L-Tyrosine.2Na.2H2O, 52.85mg/L L-Valine; Vitamins: 0.00365mg/L Biotin, 2.24mg/L D-Ca-Pantothenate, 8.98mg/L Choline chloride, 2.65mg/L Folic acid, 12.61mg/L i-Inositol, 2.0185mg/L Nicotinamide, 2.031mg/L Pyridoxine. HCl, 0.219mg/L Riboflavin, 2.17mg/L Thiamine. HCl; Other: 3151mg/L D-Glucose, 2.385mg/L Hypoxanthine, Sodium , 0.105mg/L DL-Thioctic (lipoic) acid, 0.044mg/L Methyl lineoleate, 8.1mg/L Phenol red, Sodium, 0.08mg/L Putrescine.2HCl, 110mg/L Sodium pyruvate, 0.365mg/L Thymidine.
pH 7 to 7.4
Content And Storage 2° to 8°C
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.