missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Mediatech™ Cell Culture Phosphate Buffered Saline (10X)

Description
- Maintains cell culture media in the physiological pH range
- Maintains cell tonicity and viability
- Produced under the highest industry standards to ensure superior results
- Nontoxic
- Liquid without calcium and magnesium
Specifications
Specifications
| Concentration | 10 X |
| pH | 7.3 to 7.5 |
| Physical Form | Liquid |
| Quantity | 6 x 1000 mL |
| Solution Type | Buffer |
| Formulation | Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2.40g/L, Sodium Chloride 80g/L, Disodium Phosphate 14.4g/L |
| Includes | No Calcium, No Magnesium |
| Grade | Not Available |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?