missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Phosphate Buffered Saline (1X)
BIOPHARMA

Product Code. 15373631
Change view
Click to view available options
Quantity:
6 x 1000 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15373631 6 x 1000 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15373631 Supplier Corning™ Supplier No. 21040CM
  • ONLINE EXCLUSIVE PRICE
    115.69 €
    103.18 € / Pack of 6
    Save 12.51 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With low endotoxin content. Without calcium and magnesium.

Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate. PBS helps to maintain a constant pH and is isotonic and non-toxic to cells. This formulation does not contain calcium and magnesium.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
pH 7.3 to 7.5
Physical Form Liquid
Quantity 6 x 1000 mL
Formulation Potassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.