missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Phosphate Buffered Saline (1X)
BIOPHARMA

Product Code. p-4903139
Click to view available options
Quantity:
6 x 1000 mL
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 15373631

Brand: Corning™ 21040CM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

With low endotoxin content. Without calcium and magnesium.

Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate. PBS helps to maintain a constant pH and is isotonic and non-toxic to cells. This formulation does not contain calcium and magnesium.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
pH 7.3 to 7.5
Physical Form Liquid
Quantity 6 x 1000 mL
Formulation Potassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.