Learn More
Corning™ cmRL 1066
For use in both serum supplemented and unsupplemented culture
Brand:  Corning™ 15-110-CV
Code : NEW
Additional Details : Weight : 3.76000kg
Product Code. 15343561
  • 300.79 € / Pack of 6
In Stock
to see stock.
Description
Description
(This is suggested replacement for 15-110-CVR.)
Specifications
Specifications
2° to 8°C
cmRL 1066
6.9 to 7.3
6 x 500 mL