Learn More
Corning™ Collection Bags
Product Code. 16438983 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
0.5 L
1 L
5 L
10 L
20 L
50 L
100 L
200 L
Film Description:
EVA
ULDPE
Unit Size:
Each
Product Code. Capacity (Metric) Film Description unitSize
16438983 5 L EVA Each
17283574 1 L EVA Each
17273574 0.5 L EVA Each
16458983 20 L EVA Each
16448983 10 L EVA Each
16468983 50 L EVA Each
17213584 100 L ULDPE Each
17223584 200 L ULDPE Each
8 options
Product Code. 16438983
Brand: Corning™ 9120043
Collection bags with three ports used for biopharmaceutical applications
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 L
Tubing Style Heat sealable flexible tubing
Film Description EVA
No. per Case 1/Ea.
No. per Pack 1/Pk.
Port Detail Port 1 - 0.37 in. Connector, 12 L x 0.37 I.D. x 0.62 in. O.D., chemically resistant, heat sealable flexible tubing, port 2 - male MPC connector with 0.37 in. barb, female MPC cap, port- 3 female MPC connector with 0.37 in. barb, male MPC plug, break-off i
Quantity Each