missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Clear Polypropylene 96-Well Expanded-Volume Plates
GREENER_CHOICE

Product Code. 10274972 Shop All Corning Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
10274972 Sterile Pack of 50
10091462 Non-sterile Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10274972 Supplier Corning™ Supplier No. 3344
  • ONLINE EXCLUSIVE PRICE
    399.00 €
    372.65 € / Pack of 50
    Save 26.35 €
    6% Off
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offer reduced space requirements while maintaining full instrument compatibility

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile

Order Info

Packaged in bags of ten

Certifications

Certified DNase- and RNase-free

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DNase, RNase-free
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 5 inner packs of 10
Color Clear
Product Type 96-well Assay Microplate
Material Polypropylene
Volume (Metric) Well 450 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.