missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Clear Polypropylene 96-Well Expanded-Volume Plates
Offer reduced space requirements while maintaining full instrument compatibility
311.89 € - 356.43 €
Specifications
Certifications/Compliance | DNase, RNase-free |
---|---|
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
10091462
|
Corning™
3343 |
Non-sterile |
311.89 €
Pack of 50 |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
10274972
|
Corning™
3344 |
Sterile |
356.43 €
Pack of 50 |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Order Info
Packaged in bags of ten
Certifications
Certified DNase- and RNase-free
Specifications
DNase, RNase-free | |
Without Lid | |
Clear | |
96 | |
Conical Bottom (V-well) | |
10/Pk. |
Standard | |
8 x 12 | |
Polypropylene | |
Non-treated | |
50/Cs. | |
50/Cs. |