missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Clear Polystyrene Microplates
GREENER_CHOICE

Product Code. 10134431 Shop All Corning Products
Change view
Click to view available options
Surface Treatment:
High Binding
Non-treated
Lid:
With Lid
Without Lid
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment Lid Unit Size
10134431 High Binding Without Lid Pack of 100
10481242 Non-treated Without Lid Pack of 100
10544961 Non-treated With Lid Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10134431 Supplier Corning™ Supplier No. 3700
  • ONLINE EXCLUSIVE PRICE
    885.00 €
    824.80 € / Pack of 100
    Save 60.20 €
    6% Off
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 384-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.

  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid Without Lid
    Array 16 x 24
    Binding Type High Binding
    Color Clear
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Non-sterile
    Surface Treatment High Binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.