Learn More
Corning™ 384-Well Clear Polystyrene Microplates
GREENER_CHOICE
Product Code. 10544961 Shop All Corning Products
Change view
Click to view available options
Surface Treatment:
High Binding
Non-treated
Lid:
With Lid
Without Lid
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment Lid unitSize
10544961 Non-treated With Lid Pack of 100
10134431 High Binding Without Lid Pack of 100
10481242 Non-treated Without Lid Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10544961 Supplier Corning™ Supplier No. 3680
Only null left
Add to basket
Edge
Add to basket
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 384-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid With Lid
    Culture Area 0.06 cm2
    Array 16 x 24
    Color Clear
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    No. per Case 100/Cs.
    No. per Pack 20/Pk.
    Quantity 100/Cs.