missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 96-Well, Ultra-Low Binding, Flat-Bottom Microplate
BIOPHARMAGREENER_CHOICE

Product Code. 10554961 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10554961 Clear Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10554961 Supplier Corning™ Supplier No. 3474
  • ONLINE EXCLUSIVE PRICE
    735.00 €
    683.17 € / Pack of 24
    Save 51.83 €
    7% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Color: Clear; Sterility: Sterile; Lid: With Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type Ultra-Low Attachment
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Ultra-low Binding
Total Volume (Metric) Well 360 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Maintaining cells in a suspended, unattached state
Includes Condensation rings
No. per Case 24/Cs.
No. per Pack 1/Pk.
Quantity 24/Cs.
Diameter (Metric) Well 0.64 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.