missing translation for 'onlineSavingsMsg'
Learn More

Coronene, 95%

Product Code. 10624952 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 10624952

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 191-07-1 | C24H12 | 300.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191-07-1
Molecular Formula C24H12
Molecular Weight (g/mol) 300.36
MDL Number MFCD00004134
InChI Key VPUGDVKSAQVFFS-UHFFFAOYSA-N
Synonym hexabenzobenzene, dibenzo ghi,pqr perylene, superbenzene, unii-7yy0x5xt1w, ccris 908, 7yy0x5xt1w, 6 circulene, coronene, acmc-209eul
PubChem CID 9115
ChEBI CHEBI:29863
IUPAC Name coronene
SMILES C1=CC2=CC=C3C=CC4=CC=C5C=CC6=CC=C1C1=C2C3=C4C5=C61

Specifications

Melting Point 438°C to 440°C
Color Green to Yellow
Boiling Point 525°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 500 mg
Merck Index 15, 2523
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in concentrated sulfuric acid,slightly soluble in benzene
Formula Weight 300.35
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Coronene, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Coronene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.