missing translation for 'onlineSavingsMsg'
Learn More

Cortisone acetate, MedChemExpress

Product Code. 16431866
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16431866

Brand: Medchem Express HY17461A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Cortisone acetate (Cortisone 21-acetate), an oxidized metabolite of Cortisol (a Glucocorticoid). Cortisone acetate acts as an immunosuppressant and anti-inflammatory agent. Cortisone acetate can partially intervene in binding of Glucocorticoid to Glucocorticoid-receptor at high concentrations.

Cortisone acetate (Cortisone 21-acetate), an oxidized metabolite of Cortisol (a Glucocorticoid). Cortisone acetate acts as an immunosuppressant and anti-inflammatory agent. Cortisone acetate can partially intervene in binding of Glucocorticoid to Glucocorticoid-receptor at high concentrations.

Specifications

Chemical Name or Material Cortisone acetate
Physical Form Solid
CAS 50-04-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C23H30O6
For Use With (Application) Cancer-programmed cell death
Synonym Cortisone 21-acetate
Solubility Information DMSO : 6.8 mg/mL (16.90 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (2.48 mM; ultrasonic and warming and heat to 80°C)
SMILES C[C@@]1(C2)[C@](C(COC(C)=O)=O)(O)CC[C@@]1([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])C2=O)=O
Molecular Weight (g/mol) 402.48
Formula Weight 402.48
Percent Purity 98.15%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.