missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Specifications
Specifications
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 5 inner packs of 10 |
| Binding Type | Not Treated |
| Color | Clear |
| Product Type | 96-well Assay Blocks |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
