Learn More
Corning™ 96-Well Half-Area Plates
Saves on valuable reagents by reducing the amount of reagent required per well, while retaining the ability to be read in standard microplate readers. Corning™ 96-Well Half Area Microplates are an economical solution for all of your assay diagnostic needs and are available in different colors and treatments.
Brand: Corning™ 3696
Code : 66
Additional Details : Weight : 4.03697kg
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Alphanumeric | |
With Lid | |
8 x 12 | |
TC-Treated | |
Plate readers | |
96 | |
Sterile | |
Flat Bottom (F-well) | |
50/Cs. | |
50/Cs. |
Half-Area | |
0.16 cm2 | |
Individually Wrapped | |
Clear | |
Polystyrene | |
25 to 125 μL | |
Cell Culture | |
Enhance luminescent signals and have low background luminescence and fluorescence | |
1/Pk. | |
Clear |