missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Clear Multiple Well Plates
BIOPHARMA

Product Code. 11389494 Shop All Corning Products
Change view
Click to view available options
Culture Area:
1.9 cm2
3.8 cm2
9.5 cm2
Volume (Metric) Well:
3.4 mL
6.9 mL
16.8 mL
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area Volume (Metric) Well Unit Size
11389494 9.5 cm2 16.8 mL Pack of 100
11399494 3.8 cm2 6.9 mL Pack of 100
11309504 1.9 cm2 3.4 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11389494 Supplier Corning™ Supplier No. 3736
  • ONLINE EXCLUSIVE PRICE
    320.00 €
    298.48 € / Pack of 100
    Save 21.52 €
    6% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Culture Area 9.5 cm2
Array 2 x 3
Binding Type Medium Binding
Color Clear
Material Polystyrene
Volume (Metric) Well 16.8 mL
No. of Wells 6
Volume (Metric) Working 1.9 to 2.9 mL
Sterility Sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 5/Pk.
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.