missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Clear Multiple Well Plates
BIOPHARMA

Product Code. 11399494 Shop All Corning Products
Change view
Click to view available options
Culture Area:
1.9 cm2
3.8 cm2
9.5 cm2
Volume (Metric) Well:
3.4 mL
6.9 mL
16.8 mL
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area Volume (Metric) Well Unit Size
11399494 3.8 cm2 6.9 mL Pack of 100
11309504 1.9 cm2 3.4 mL Pack of 100
11389494 9.5 cm2 16.8 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399494 Supplier Corning™ Supplier No. 3737
  • ONLINE EXCLUSIVE PRICE
    361.00 €
    337.22 € / Pack of 100
    Save 23.78 €
    6% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 14-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Bulk Quote Request

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Spezifikation

Format Standard
Lid With Lid
Culture Area 3.8 cm2
Array 3 x 4
Binding Type Medium Binding
Color Clear
Material Polystyrene
Volume (Metric) Well 6.9 mL
No. of Wells 12
Volume (Metric) Working 0.76 to 1.14 mL
Sterility Sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 5/Pk.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.