missing translation for 'onlineSavingsMsg'
Learn More

Corning™ HTS Transwell™ -96 Well Permeable Support System
BIOPHARMA

Product Code. 10555673 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
No. per Pack:
1/Pk.
4/Pk.
5/Pk.
Unit Size:
Each
Pack of 2
Pack of 5
Pack of 8
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size No. per Pack Unit Size
10555673 5 μm 1/Pk. Pack of 2
10175562 0.4 μm 5/Pk. Pack of 5
10625053 0.4 μm 1/Pk. Each
10399242 1 μm 5/Pk. Pack of 5
10043832 1 μm 1/Pk. Each
10271593 3 μm 1/Pk. Pack of 2
10077792 3 μm 4/Pk. Pack of 8
10797023 5 μm 4/Pk. Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10555673 Supplier Corning™ Supplier No. 3388
  • ONLINE EXCLUSIVE PRICE
    215.00 €
    193.97 € / Pack of 2
    Save 21.03 €
    9% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Boasts membrane area for cell growth or migration of 0.143cm2. Corning™ HTS Transwell™ 96-Well Permeable Support System is a microplate support system for an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.

  • The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
  • Choice of either polyester (PET) membrane (0.4 μm, 1.0 μm pore sizes) or polycarbonate (PC) membrane (0.4 μm, 3.0 μm, and 5.0 μm pore sizes)
  • 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
  • Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access.
  • Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
  • The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
  • The receiver plate isolates each well to enable 96 individual assays.
  • Sterile
  • The HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid.
  • The HTS Transwell-96 plates (3.0 and 5.0 μm PC, 8.0 μm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Culture Area 0.143 cm2
Membrane TC-treated polycarbonate
Array 8 x 12
Pore Size 5 μm
Color Clear
Material Polycarbonate
No. of Wells 96
Volume (Metric) Working 0.23 mL
Sterility Sterile
Well Shape Flat Bottom (F-well)
Includes HTS Transwell™-96 permeable support insert, receiver plate (96 wells for growth or assay steps) and 1 sterile lid
No. per Case 2/Cs.
No. per Pack 1/Pk.
Diameter (Metric) Membrane 4.26 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.