missing translation for 'onlineSavingsMsg'
Learn More

Coumarin-3-carbonitrile, 97%

Product Code. 11387489
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387489 5 g 5g
11377489 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387489 Supplier Thermo Scientific Alfa Aesar Supplier No. H27871.06
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15119-34-3 | C10H5NO2 | 171.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15119-34-3
Molecular Formula C10H5NO2
Molecular Weight (g/mol) 171.16
MDL Number MFCD00115699
InChI Key QKJALQPLNMEDAV-UHFFFAOYSA-N
Synonym 3-cyanocoumarin, 2-oxo-2h-chromene-3-carbonitrile, coumarin-3-carbonitrile, 2h-1-benzopyran-3-carbonitrile, 2-oxo, 2-oxo-2h-1-benzopyran-3-carbonitrile, coumarin, 3-cyano-7ci,8ci, 3-cyano-coumarin, pubchem8675, acmc-1cfdq
PubChem CID 203763
IUPAC Name 2-oxochromene-3-carbonitrile
SMILES O=C1OC2=CC=CC=C2C=C1C#N

Specifications

Melting Point 184°C to 188°C
Quantity 5 g
Formula Weight 171.16
Percent Purity 97%
Chemical Name or Material Coumarin-3-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.