missing translation for 'onlineSavingsMsg'
Learn More

Coumarin, 98%

Product Code. 11458246
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458246 5000 g 5000g
11448246 250 g 250g
11438246 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458246 Supplier Thermo Scientific Alfa Aesar Supplier No. A15336.0I
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-64-5 | C9H6O2 | 146.15 g/mol

Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-64-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006850
InChI Key ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
Synonym coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone
PubChem CID 323
ChEBI CHEBI:28794
SMILES O=C1OC2=CC=CC=C2C=C1

Specifications

Melting Point 68°C to 71°C
Density 0.935
Boiling Point 297°C to 299°C
Flash Point 162°C (323°F)
Odor Pleasant
Quantity 5000 g
UN Number UN2811
Beilstein 383644
Merck Index 14,2562
Solubility Information Soluble in water.
IUPAC Name chromen-2-one
Formula Weight 146.15
Percent Purity 98%
Chemical Name or Material Coumarin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.