missing translation for 'onlineSavingsMsg'
Learn More

Coumatetralyl, TRC

Product Code. 30041101
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041101 250 mg 250mg
30133706 100 mg 100mg
30040716 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041101 Supplier Toronto Research Chemicals Supplier No. TRCC765600250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5836-29-3
Molecular Formula C19 H16 O3
Molecular Weight (g/mol) 292.33
IUPAC Name 4-hydroxy-3-(1,2,3,4-tetrahydronaphthalen-1-yl)chromen-2-one
SMILES OC1=C(C2CCCc3ccccc23)C(=O)Oc4ccccc14

Specifications

Quantity 250 mg
Synonym Coumarin, 4-hydroxy-3-(1,2,3,4-tetrahydro-1-naphthyl)- (6CI,7CI,8CI), 4-Hydroxy-3-(1,2,3,4-tetrahydro-1-naphthalenyl)-2H-1-benzopyran-2-one, 3-(1,2,3,4-Tetrahydro-1-naphthyl)-4-hydroxycoumarin, 3-(α-Tetralinyl)-4-hydroxycoumarin, 3-(α-Tetralyl)-4-Hydroxycoumarin, 4-Hydroxy-3-(1,2,3,4-tetrahydro-1-naphthyl)coumarin, Coumatetralyl, Endrocide, Murisan A, Racumin, Racumin 57, Rodentin
InChI Formula InChI=1S/C19H16O3/c20-18-15-9-3-4-11-16(15)22-19(21)17(18)14-10-5-7-12-6-1-2-8-13(12)14/h1-4,6,8-9,11,14,20H,5,7,10H2
Formula Weight 292.1099
Chemical Name or Material Coumatetralyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.