Learn More
Macherey-Nagel™ Cover Lids for NucleoFast™ 96 PCR Plates
Product Code. 15850978
Change view
Click to view available options
:
50 Lids
Product Code.
15850978 50 Lids
1 options
Product Code. 15850978 Supplier Macherey-Nagel™ Supplier No. 743101.50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
To prevent contamination and spills when working with NucleoFast™ 96-well ultrafiltration plates for PCR clean-up.
NucleoFast™ 96 PCR
  • Cost-efficient 96-well ultrafiltration kit for PCR clean-up
  • Ready-to-use DNA for sequencing and microarray spotting
  • Sturdy membrane allows easy recovery of DNA
  • No well-to-well cross-contamination
  • Fast procedure
  • Available as complete kits and as plates only
  • Detergent-free membrane
  • Manual or automated use