Learn More
CleanSpace™ Technology Coverall
Product Code. 15940062 Shop All CleanSpace Technology Products
Change view
Click to view available options
Quantity:
25/Pk.
Unit Size:
Each
Product Code. Quantity Unit Size
15940062 25/Pk. Each
1 options
Product Code. 15940062 Supplier CleanSpace™ Technology Supplier No. PAF0049
  • ONLINE EXCLUSIVE PRICE
    62.20 €
    48.00 € / Pack of 1
    Save 14.20 €
    22% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Disposable coverall protects CleanSpace™ respirators from dirt and splash.
  • Used with the revolutionary CleanSpace™- a light weight PAPR with no hoses/belts
  • Materials: Spun polymer fibres
  • Easy and quickly fitted and removed from the power unit
  • Compatible with all CleanSpace™ Respirators
Specifications
Disposable Reusable
Product Type Blower Cover
For Use With (Equipment) Standard and Large Case Filters
Model PAF-0049
Quantity 25/Pk.