Learn More
COX4 Antibody (CL3501) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59777UV
Product Code. 30293643
  • 737.00 € / 0.10mL
Description
COX4 Monoclonal antibody specifically detects COX4 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COX4
Monoclonal
DyLight 350
COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016
This antibody was developed using a recombinant protein derived from P13073, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Loading Controls, Neuroendocrinology
1327
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL3501
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified