missing translation for 'onlineSavingsMsg'
Learn More

CP-547632, MedChemExpress

Product Code. 16468327
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468327 Supplier Medchem Express Supplier No. HY13302
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CP-547632 is an orally active, ATP-competitive and potent VEGFR-2 and FGF kinases inhibitor with IC50s of 11 nM and 9 nM, respectively. CP-547632 is selective for VEGFR2 and bFGF over EGFR, PDGFRβ, and related tyrosine kinases (TKs). CP-547632 has antitumor efficacy.

CP-547632 is an orally active, ATP-competitive and potent VEGFR-2 and FGF kinases inhibitor with IC50s of 11 nM and 9 nM, respectively. CP-547632 is selective for VEGFR2 and bFGF over EGFR, PDGFRβ, and related tyrosine kinases (TKs). CP-547632 has antitumor efficacy.

Specifications

Chemical Name or Material CP-547632
CAS 252003-65-9
Color White
Purity Grade Notes Research
Molecular Formula C20H24BrF2N5O3S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (187.83 mM; Need ultrasonic)
SMILES O=C(NC1=C(C(N)=O)C(OCC2=C(F)C=C(Br)C=C2F)=NS1)NCCCCN3CCCC3
Molecular Weight (g/mol) 532.4
Formula Weight 532.4
Percent Purity 98.01%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.