missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPEB1 Antibody (SR2146), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | CPEB1 |
|---|---|
| Clone | SR2146 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676654
|
Novus Biologicals
NBP3-22113-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694314
|
Novus Biologicals
NBP3-22113-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPEB1 Monoclonal antibody specifically detects CPEB1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| CPEB1 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CPEBCEBP, CPE-binding protein 1, CPE-BP1, cytoplasmic polyadenylation element binding protein 1, cytoplasmic polyadenylation element-binding protein 1, FLJ13203, h-CEBP, hCPEB-1, MGC34136, MGC60106 | |
| A synthesized peptide derived from human CPEB1 (Uniprot #: Q9BZB8) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2146 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 64506 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title