missing translation for 'onlineSavingsMsg'
Learn More

CpG oligodeoxynucleotides (2216), Novus Biologicals™

Product Code. 18154912 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18154912 0.025 mg 25µg
18015644 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18154912 Supplier Novus Biologicals Supplier No. NBP231136SS
  • ONLINE EXCLUSIVE PRICE
    194.00 €
    182.70 € / 25µg
    Save 11.30 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Useful positive control model system for studying hTLR9 activation

Immunogen: G&asterisk;GGGGACGATCGTCG&asterisk;G&asterisk;G&asterisk;G&asterisk;G&asterisk;G Buffer: Sterile water Clone: 2216 Dilution: Ligand Activation, 5 to 20μg/mL CpG ODN Type A (2216)
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.0 mg/mL
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.025 mg
For Use With (Application) Human CpG ODNs are used for activation of human TLR9 at a range of 5 to 20μL. The TLR9/NF-kB/SEAPorter HEK 293 cell line is a useful positive control model system for studying hTLR9 activation. Typically use a 96-well plate format assay using this reporter cell line
Type CpG Oligodeoxynucleotides
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.