missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF4L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92334-0.02ml
This item is not returnable.
View return policy
Description
CPSF4L Polyclonal antibody specifically detects CPSF4L in Mouse samples. It is validated for Western Blot
Specifications
| CPSF4L | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| cleavage and polyadenylation specific factor 4-like, putative cleavage and polyadenylation specificity factor subunit 4-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-228 of mouse Cpsf4l (NP_080958.2). MEEVIAGLQGFTFAFELDVESQKGTGLLPFQGMDKSSSAVCNFFAKGLCVKGMLCPLRHEQGEKLVVCKHWLRGLCRKSDCCDFLHQYDVSKMPVCYFHSKFGNCSNKECLFLHLKPVLKLQDCPWYNQGFCKEVGPLCKYRHVHQVLCPNYFTGFCPEGPQCQFGHPKMSPPFHPSNVKLQPVNQPWDSTTSTGNTLVSPFQAKPMVHGQRRWSLPQACSSRAHPAP | |
| 0.02 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 642843 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction