missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPT1C Recombinant Rabbit Monoclonal Antibody (6N7B2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558197
This item is not returnable.
View return policy
Description
Immunogen sequence: MAEAHQAVGF RPSLTSDGAE VELSAPVLQE IYLSGLRSWK RHLSRFWNDF.
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.
Specifications
| CPT1C | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| F1LN46, Q8BGD5 | |
| CPT1C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-50 of human CPT1C (NP_689572.1). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 6N7B2 | |
| Unconjugated | |
| CPT1C | |
| 6530437J22Rik; 9630004I06Rik; Carnitine O-palmitoyltransferase 1, brain isoform; carnitine O-palmitoyltransferase I, brain isoform; carnitine palmitoyltransferase 1, brain; carnitine palmitoyltransferase 1c; carnitine palmitoyltransferase I related C; CATL1; CPT IC; CPT I-C; CPT1-B; Cpt1c; CPT1P; CPTI-B; CPTIC; FLJ23809; SPG73 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 308579, 78070 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction