missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85470
This item is not returnable.
View return policy
Description
CPT2 Polyclonal antibody specifically detects CPT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CPT2 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| carnitine O-palmitoyltransferase 2, mitochondrial, carnitine palmitoyltransferase 2, Carnitine palmitoyltransferase IIEC 2.3.1.21, CPT II, CPT1, CPTASE | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MMVECSKYHGQLTKEAAMGQGFDRHLFALRHLAAAKGIILPELYLDPAYGQINHNVLSTSTLSSPAVNLGGFAPVVSDGFGVGYAVHDNWIGCNVSSYP | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 1376 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction