Learn More
Invitrogen™ CRABP1 Recombinant Rabbit Monoclonal Antibody (23GB5105), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552792
 View more versions of this product
Product Code. 30089150
  • 446.00 € / 100µL
Description
This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CRABP1
Recombinant Monoclonal
Unconjugated
CRABP1
AI326249; cCRABP-I; cellular retinoic acid binding protein 1; cellular retinoic acid binding protein I; cellular retinoic acid binding protein I (CRABP-I) homologue; cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; cellular retinoic-acid-binding protein; CRABP; Crabp1; Crabp-1; CrabpI; CRABP-I; RBP5; Rbp-5; retinoic acid-binding protein
Rabbit
Affinity chromatography
RUO
12903, 1381
-20°C
Liquid
Western Blot, Immunocytochemistry
23GB5105
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P29762, P62965
CRABP1
A synthesized peptide derived from human CRABP1 (100-137AA).
100 μL
Primary
Human, Mouse, Rat