missing translation for 'onlineSavingsMsg'
Learn More

Crackle Test Reference Standard: 0 wt% H2O in 10W30 Motor Oil, VHG™

Product Code. 30002926
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 30002926

Brand: VHG Labs VHGCTRBLK100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Crackle Test Reference Standard: 0 wt% H2O in 10W30 Motor Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Water
CAS 7732-18-5
Analyte or Component Names Water
Shipping Condition Room Temperature
Molecular Formula H2 O
Synonym Water, Dihydrogen oxide
InChI Formula InChI=1S/H2O/h1H2
SMILES O
IUPAC Name oxidane
Solvent or Matrix 10W30 Motor Oil
Molecular Weight (g/mol) 18.02
Quantity 100 mL
Formula Weight 18.0106 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.