Learn More
Edmund Bühler GmbH™ Cramp Strip
Product Code. 10023511
Click to view available options
For Use With (Equipment):
Combifix
KL and KS shakers
SM Shakers
Unit Size:
Each
Product Code. 10023511
Brand: Edmund Bühler GmbH™ 0050 401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For fastening horizontal separating funnels. Includes clamps for NS 19 and NS 29.
  • Keeps horizontal separating funnels in place
  • Includes clamps for NS 19 and NS 29
Specifications
Product Type Cramp Strip
For Use With (Application) For fastening horizontal separating funnels
For Use With (Equipment) SM Shakers
Includes 5 Clamps for NS 19, 2 clamps for NS 29
No. of Positions 7