missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRCP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92783-0.02ml
This item is not returnable.
View return policy
Description
CRCP Polyclonal antibody specifically detects CRCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CRCP | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Calcitonin gene-related peptide-receptor component protein, CGRP receptor component, CGRPRCP, CGRP-RCPRNA polymerase III subunit C9, CGRP-receptor component protein, DNA-directed RNA polymerase III subunit RPC9, hsC17, RCP9, RCPMGC111194 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human CRCP (NP_001035737.1). MEVKDANSALLSNYETLKYISKTPCRHQSPEIVREFLTALKSHKLTKAEKLQLLNHRPVTAVEIQLMVEESEERLTEEQIEALLHTVTSILPAEPEAEQKKNTNSNVAMDEEDPA | |
| 0.02 mL | |
| GPCR, Neuroscience, Signal Transduction | |
| 27297 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction