missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cre Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56134-0.025ml
This item is not returnable.
View return policy
Description
Cre Polyclonal antibody specifically detects Cre in Mouse, Cre/lox System samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page, Immunohistochemistry Free-Floating
Specifications
| Cre | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000. Use reported in scientific literature (PMID 33784255), Immunohistochemistry 1:10 - 1:5000, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:10 - 1:5000, SDS-Page reported in scientific literature (PMID 27432829), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25429143) | |
| Cyclization recombinase | |
| A synthethic peptide corresponding to a portion of amino acids 1-50 of the bacteriophage protein CRE was used as immunogen, GenBank no. AAQ14086.1. | |
| 0.025 mL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| 50 uL neat whole antisera | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Mouse, Cre/lox System | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction