missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase BB Antibody (CKB/1268), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75762-100ug
This item is not returnable.
View return policy
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for ELISA, Protein Array
Specifications
| Creatine Kinase BB | |
| Monoclonal | |
| 0.2 mg/mL | |
| 10 mM PBS with 0.05% BSA | |
| Creatine Kinase Isoenzyme | |
| Recombinant full-length human Creatine Kinase BB protein (Uniprot: P12277) | |
| 100 μg | |
| Cancer | |
| 1152 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA, Protein Array | |
| CKB/1268 | |
| Unconjugated | |
| P12277, P12277, P12277, P12277 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction