missing translation for 'onlineSavingsMsg'
Learn More

CREM Monoclonal Antibody (OTI4H9), TrueMAB™, OriGene

Product Code. p-200013302
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17566324 30 μL 30µL
17546324 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17566324 Supplier Origene Technologies Supplier No. TA812012S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CREM Monoclonal antibody specifically detects CREM in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen CREM
Applications Western Blot
Classification Monoclonal
Clone OTI4H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Klf7
Gene Accession No. O75840, Q99JB0
Gene Alias cAMP response element modulator; cAMP responsive element moderator; cAMP responsive element modulator; cAMP-responsive element modulator; CRE modulator; CREM; CREM 2alpha-b protein; CREM 2beta-a protein; CREM-2; hCREM-2; ICER; ICERI; inducible cAMP early repressor; inducible cAMP early repressor ICER; inducible cyclic AMP early repressor; MGC111110; MGC17881; MGC41893; OTTHUMP00000019443
Host Species Mouse
Immunogen Full length human recombinant protein of human KLF7 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8609, 93691
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.