missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Cresyl Violet acetate

Product Code. 16444716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444716 Supplier MedChemExpress Supplier No. HY101888
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cresyl Violet acetate is a red fluorescent stain, which can be used to stain neurons.

Specifications

Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : 1 mg/mL (3.11 mM; Need ultrasonic) ∣ H2O : 0.67 mg/mL (2.09 mM; Need ultrasonic)
Molecular Weight (g/mol) 321.33 g/mol
CAS 10510-54-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.