Learn More
CRISPR-Cas9 Antibody (7A9-3A3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36440H
This item is not returnable.
View return policy
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Mouse, Bacteria samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount, Knockout Validated.
Specifications
| Immunohistochemistry (Frozen), Immunoassay | |
| Protein G purified | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Monoclonal | |
| This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus is specific toCas9 protein from Streptococcus pyogene serotype M1. | |
| Purified |