missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80679
This item is not returnable.
View return policy
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Mouse, Bacteria samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, Knockout Validated.
Specifications
| CRISPR-Cas9 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:1000, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 1:500, Immunoprecipitation, Immunohistochemistry-Frozen reported in scientific literature (PMID 28153089), Immunohistochemistry Whole-Mount, Knockout Validated | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2]. | |
| 0.1 mL | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| 901176 | |
| Mouse, Bacteria | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay | |
| 7A9-3A3 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse reactivity reported in multiple pieces of scientific literature. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction