missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CrkL Antibody (S02-3D8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19852-50ul
This item is not returnable.
View return policy
Description
CrkL Monoclonal antibody specifically detects CrkL in Human samples. It is validated for Western Blot
Specifications
| CrkL | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S02-3D8 | |
| Unconjugated | |
| crk-like protein, pCRKL, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like | |
| A synthetic peptide of human CrkL (Uniprot # P46109) | |
| 50 μg | |
| Cancer, Phospho Specific, Signal Transduction | |
| 1399 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction